Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207385.1 | NP_997268.1 | 3 | 332 | ||
XM_017023228.1 | XP_016878717.1 | 8 | 2014 | |||
XM_017023229.1 | XP_016878718.1 | 3 | 620 | |||
GenBank mRNA | AK129695.1 | - | 3 | 332 |