Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167985.1 | NP_001161457.1 | 4 | 417 | ||
NM_001167986.1 | NP_001161458.1 | 5 | 704 | |||
NM_001167987.1 | NP_001161459.1 | 4 | 417 | |||
NM_213726.2 | NP_998891.2 | 4 | 579 | |||
XM_005256627.3 | XP_005256684.1 | 3 | 612 | |||
XM_005256628.4 | XP_005256685.1 | 3 | 809 | |||
XM_005256629.1 | XP_005256686.1 | 3 | 664 | |||
XM_006721517.1 | XP_006721580.1 | 3 | 664 | |||
XM_006721518.2 | XP_006721581.1 | 3 | 613 | |||
XM_006721519.3 | XP_006721582.1 | 3 | 810 | |||
XM_011523832.2 | XP_011522134.1 | 4 | 934 | |||
XM_011523834.2 | XP_011522136.1 | 3 | 612 | |||
GenBank mRNA | AY601906.1 | - | 4 | 579 | ||
AY601907.1 | - | 4 | 417 | |||
AY604176.1 | - | 2 | 137 | |||
BC119781.1 | - | 4 | 493 | 40109268 | ||
BC122873.1 | - | 3 | 206 | 40109270 |