Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204118.1 | NP_001191047.1 | 12 | 920 | ||
NM_207390.3 | NP_997273.3 | 12 | 920 | |||
NR_109784.1 | 12 | 983 | ||||
XM_017026785.1 | XP_016882274.1 | 13 | 1464 | |||
XM_017026786.1 | XP_016882275.1 | 13 | 1328 | |||
XM_017026787.1 | XP_016882276.1 | 14 | 1683 | |||
XM_017026788.1 | XP_016882277.1 | 13 | 1440 | |||
XM_017026789.1 | XP_016882278.1 | 9 | 1142 | |||
XM_017026790.1 | XP_016882279.1 | 12 | 1416 | |||
XM_017026791.1 | XP_016882280.1 | 11 | 1265 | |||
XM_017026792.1 | XP_016882281.1 | 13 | 1468 | |||
GenBank mRNA | BC140848.1 | - | 12 | 881 | 9021716 | |
BC144664.1 | - | 12 | 944 |
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832663.1 | - | 16 | 2197 |