Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242800.1 | NP_001229729.1 | 5 | 454 | ||
NM_001242801.1 | NP_001229730.1 | 5 | 632 | |||
NM_001242802.1 | NP_001229731.1 | 5 | 722 | |||
NM_206894.3 | NP_996777.2 | 5 | 414 | |||
XM_005258903.4 | XP_005258960.1 | 5 | 781 | |||
XM_011526950.2 | XP_011525252.1 | 5 | 862 | |||
GenBank mRNA | AK297474.1 | - | 5 | 423 | ||
BC057245.1 | - | 5 | 416 | 6712908 | ||
BM473790.1 | - | 3 | 254 | |||
DA819130.1 | - | 5 | 412 | |||
DC308617.1 | - | 5 | 456 | |||
EL735861.1 | - | 3 | 293 |