Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006605.4 | NP_001006606.2 | 4 | 520 | ||
NM_001252269.1 | NP_001239198.1 | 3 | 385 | |||
NM_001252271.1 | NP_001239200.1 | 4 | 563 | |||
NM_001252273.1 | NP_001239202.1 | 4 | 520 | |||
GenBank mRNA | AK027146.1 | - | 1 | 81 | ||
AK303700.1 | - | 3 | 538 | |||
AL536469.3 | - | 4 | 464 | |||
AL702320.1 | - | 4 | 484 | |||
BC070342.1 | - | 4 | 454 | 6498824 | ||
DA141589.1 | - | 4 | 533 | |||
DA155419.1 | - | 4 | 437 | |||
DA736826.1 | - | 3 | 261 | |||
DB199113.1 | - | 3 | 311 |