Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 1 | 863 | ||
XM_011509963.2 | XP_011508265.1 | 1 | 880 | |||
GenBank mRNA | AB096683.1 | - | 1 | 863 | ||
AK307197.1 | - | 1 | 839 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 1 | 880 | ||
XM_011509724.2 | XP_011508026.1 | 1 | 880 |