Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 10 | 1133 | ||
NM_001287191.1 | NP_001274120.1 | 9 | 1037 | |||
NM_001287192.1 | NP_001274121.1 | 9 | 1037 | |||
XM_005252254.2 | XP_005252311.1 | 10 | 1366 | |||
XM_005252257.3 | XP_005252314.1 | 9 | 1032 | |||
XM_005252260.3 | XP_005252317.1 | 8 | 999 | |||
XM_011519077.2 | XP_011517379.1 | 11 | 1024 | |||
XM_011519078.2 | XP_011517380.2 | 10 | 1031 | |||
XM_011519079.2 | XP_011517381.2 | 10 | 1033 | |||
XM_011519080.2 | XP_011517382.1 | 4 | 1729 | |||
XM_011519081.2 | XP_011517383.1 | 4 | 1714 | |||
XM_017015182.1 | XP_016870671.1 | 10 | 1366 | |||
XM_017015183.1 | XP_016870672.1 | 9 | 1032 | |||
XM_017015184.1 | XP_016870673.1 | 5 | 542 | |||
XM_017015185.1 | XP_016870674.1 | 10 | 1039 | |||
XM_017015186.1 | XP_016870675.1 | 4 | 382 | |||
XM_017015187.1 | XP_016870676.1 | 9 | 1032 | |||
GenBank mRNA | AK296433.1 | - | 9 | 948 | ||
AK299350.1 | - | 10 | 1065 | |||
AK299491.1 | - | 9 | 948 | |||
AK309817.1 | - | 10 | 1374 | |||
BC036746.1 | - | 4 | 476 | 4384472 |