Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 12 | 1361 | ||
NM_001287191.1 | NP_001274120.1 | 12 | 1295 | |||
NM_001287192.1 | NP_001274121.1 | 11 | 1262 | |||
XM_005252254.2 | XP_005252311.1 | 13 | 1627 | |||
XM_005252257.3 | XP_005252314.1 | 12 | 1293 | |||
XM_005252260.3 | XP_005252317.1 | 11 | 1260 | |||
XM_011519077.2 | XP_011517379.1 | 14 | 1285 | |||
XM_011519078.2 | XP_011517380.2 | 12 | 1259 | |||
XM_011519079.2 | XP_011517381.2 | 12 | 1258 | |||
XM_011519080.2 | XP_011517382.1 | 7 | 1990 | |||
XM_011519081.2 | XP_011517383.1 | 6 | 1942 | |||
XM_017015182.1 | XP_016870671.1 | 12 | 1591 | |||
XM_017015183.1 | XP_016870672.1 | 11 | 1260 | |||
XM_017015184.1 | XP_016870673.1 | 8 | 803 | |||
XM_017015186.1 | XP_016870675.1 | 7 | 643 | |||
GenBank mRNA | AK296433.1 | - | 12 | 1206 | ||
AK299350.1 | - | 12 | 1293 | |||
AK299491.1 | - | 11 | 1173 | |||
BC036746.1 | - | 6 | 704 | 4384472 | ||
CR936789.1 | - | 3 | 353 |