Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVFRD-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207582.2 | NP_997465.1 | 2 | 1594 | ||
GenBank mRNA | AK123938.1 | - | 2 | 1479 | ||
AY358244.1 | - | 1 | 1222 | |||
BC068585.1 | - | 2 | 1482 | 30346522 |