Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281765.2 | NP_001268694.1 | 18 | 3792 | ||
NM_001281766.2 | NP_001268695.1 | 17 | 3726 | |||
NM_001318761.1 | NP_001305690.1 | 18 | 2910 | |||
NM_004439.7 | NP_004430.4 | 18 | 3789 | |||
NM_182472.4 | NP_872272.2 | 17 | 3723 | |||
XM_005265653.3 | XP_005265710.1 | 16 | 3643 | |||
XM_011531735.2 | XP_011530037.1 | 17 | 3799 | |||
XM_017007878.1 | XP_016863367.1 | 16 | 3733 | |||
XM_017007880.1 | XP_016863369.1 | 15 | 3577 | |||
GenBank mRNA | BC136257.1 | - | 18 | 3059 | 9020244 | |
BC136258.1 | - | 18 | 3059 | 9020245 | ||
BC143427.1 | - | 17 | 2993 | 9051935 | ||
BX537946.1 | - | 18 | 2911 | |||
L36644.1 | - | 17 | 2898 |