Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012418.3 | NP_001012418.2 | 9 | 1079 | ||
XM_005249078.3 | XP_005249135.2 | 9 | 1044 | |||
XM_005249079.2 | XP_005249136.1 | 9 | 937 | |||
XM_006715082.3 | XP_006715145.1 | 8 | 2374 | |||
XM_011514567.2 | XP_011512869.1 | 9 | 1167 | |||
XM_017010819.1 | XP_016866308.1 | 9 | 976 | |||
GenBank mRNA | AK122581.1 | - | 1 | 83 | ||
BC132831.1 | - | 8 | 845 | 40146853 | ||
BC132833.1 | - | 8 | 845 | 40146855 | ||
BX647582.1 | - | 9 | 1079 |