Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 21 | 1990 | ||
XM_011535066.2 | XP_011533368.1 | 20 | 1996 | |||
XM_017020555.1 | XP_016876044.1 | 20 | 1998 | |||
XM_017020556.1 | XP_016876045.1 | 20 | 2002 | |||
XM_017020557.1 | XP_016876046.1 | 21 | 2081 | |||
GenBank mRNA | BC063885.1 | - | 21 | 1976 | 30383658 |