Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005861.2 | NP_001005861.1 | 13 | 1632 | ||
NM_002958.3 | NP_002949.2 | 13 | 1623 | |||
XM_017007016.1 | XP_016862505.1 | 8 | 1129 | |||
GenBank mRNA | AB209405.1 | - | 3 | 3959 | ||
BC021700.2 | - | 8 | 898 | |||
S59184.1 | - | 13 | 1656 | |||
X69970.1 | - | 13 | 1623 | |||
X96588.1 | - | 14 | 1528 |