Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004197.1 | NP_004188.1 | 2 | 413 | ||
NM_032454.1 | NP_115830.1 | 2 | 413 | |||
NR_026717.1 | 2 | 598 | ||||
GenBank mRNA | AK302111.1 | - | 2 | 612 | ||
AK303438.1 | - | 2 | 451 | |||
BC110347.1 | - | 2 | 598 | 5165123 | ||
BC121128.1 | - | 1 | 432 | 40120738 | ||
BC121129.1 | - | 1 | 431 | 40120740 | ||
BC143833.1 | - | 1 | 468 | 9052348 | ||
BC143835.1 | - | 1 | 468 | |||
L26260.1 | - | 2 | 413 |