Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286074.1 | NP_001273003.1 | 36, 37 | 5207 | ||
NM_004606.4 | NP_004597.2 | 35, 36 | 5201 | |||
NM_138923.3 | NP_620278.1 | 35, 36 | 5138 | |||
NR_104389.1 | 35, 36 | 5201 | ||||
NR_104390.1 | 36, 37 | 5207 | ||||
NR_104391.1 | 37, 38 | 5229 | ||||
NR_104392.1 | 36, 37 | 5223 | ||||
NR_104394.1 | 36, 37 | 5180 | ||||
NR_104395.1 | 36, 37 | 5232 | ||||
GenBank mRNA | AB300418.1 | - | 36, 37 | 5134 | ||
AJ549250.3 | - | 10, 11 | 1026 | |||
AJ555148.2 | - | 11, 12 | 1032 | |||
AJ555149.2 | - | 12, 13 | 1054 | |||
AM711892.1 | - | 11, 12 | 1048 | |||
AM711894.1 | - | 11, 12 | 1005 | |||
AM711895.1 | - | 11, 12 | 1057 | |||
D90359.1 | - | 35, 36 | 5113 | |||
M73444.1 | - | 5, 6 | 486 |