Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 5 | 540 | ||
NM_001324236.1 | NP_001311165.1 | 6 | 666 | |||
NM_001324237.1 | NP_001311166.1 | 7 | 571 | |||
NM_001324238.1 | NP_001311167.1 | 6 | 574 | |||
NM_033453.3 | NP_258412.1 | 7 | 663 | |||
NM_181493.3 | NP_852470.1 | 7 | 612 | |||
NR_052000.1 | 6 | 506 | ||||
NR_052001.2 | 6 | 584 | ||||
NR_052002.1 | 6 | 598 | ||||
XM_006723564.3 | XP_006723627.1 | 8 | 993 | |||
XM_006723565.3 | XP_006723628.1 | 6 | 870 | |||
GenBank mRNA | AB062127.1 | - | 7 | 532 | ||
AF026816.2 | - | 7 | 522 | |||
AF219116.1 | - | 7 | 575 | |||
AF334680.1 | - | 6 | 455 | |||
BC010138.2 | - | 7 | 507 | 4124624 | ||
BE275997.1 | - | 6 | 541 | |||
BF969192.1 | - | 6 | 506 | |||
BI115811.1 | - | 5 | 376 | |||
BM560812.1 | - | 663 | ||||
BQ049090.1 | - | 6 | 445 | |||
CA425905.1 | - | 2 | 536 | |||
EF199841.1 | - | 7 | 510 | |||
EF213026.1 | - | 7 | 459 | |||
KU178148.1 | - | 7 | 471 | |||
KU178149.1 | - | 5 | 348 |