Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007271.1 | NP_001007272.1 | 8 | 1391 | ||
NM_001007272.1 | NP_001007273.1 | 4 | 666 | |||
NM_001007273.1 | NP_001007274.1 | 6 | 965 | |||
NM_001320842.1 | NP_001307771.1 | 7 | 1183 | |||
NM_001320843.1 | NP_001307772.1 | 5 | 761 | |||
NM_016364.3 | NP_057448.3 | 4 | 524 | |||
XM_005269883.3 | XP_005269940.1 | 7 | 1161 | |||
XM_005269884.4 | XP_005269941.3 | 4 | 953 | |||
XM_005269887.1 | XP_005269944.1 | 7 | 1263 | |||
XM_005269890.1 | XP_005269947.1 | 5 | 863 | |||
XM_011539853.1 | XP_011538155.1 | 8 | 1400 | |||
XM_011539854.2 | XP_011538156.1 | 6 | 939 | |||
XM_011539855.1 | XP_011538157.1 | 4 | 642 | |||
XM_011539856.2 | XP_011538158.1 | 6 | 943 | |||
XM_017016313.1 | XP_016871802.1 | 4 | 616 | |||
GenBank mRNA | AB027004.1 | - | 4 | 538 | ||
AB103375.1 | - | 4 | 538 | |||
AI971328.1 | - | 1 | 394 | |||
AK127886.1 | - | 7 | 1183 | |||
AY674051.1 | - | 8 | 1364 |