Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159488.1 | NP_001152960.1 | 6 | 1198 | ||
NM_030764.3 | NP_110391.2 | 6 | 1198 | |||
XM_011509974.2 | XP_011508276.1 | 6 | 1533 | |||
XM_011509975.2 | XP_011508277.1 | 6 | 1535 | |||
XM_011509976.2 | XP_011508278.1 | 6 | 1534 | |||
XM_017002316.1 | XP_016857805.1 | 6 | 1533 | |||
XM_017002317.1 | XP_016857806.1 | 6 | 1336 | |||
XM_017002319.1 | XP_016857808.1 | 5 | 1248 | |||
GenBank mRNA | AF319438.1 | - | 1 | 683 | ||
AF319439.1 | - | 1 | 521 | |||
AF319440.1 | - | 1 | 671 | |||
AF390037.1 | - | 1 | 521 | |||
AF459633.1 | - | 6 | 1186 | |||
AK301113.1 | - | 6 | 1185 | |||
AL833194.1 | - | 1 | 3057 | |||
AY043465.1 | - | 6 | 1185 | |||
AY358130.1 | - | 6 | 1188 | |||
BC129836.1 | - | 1 | 523 | 40115446 |