Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080426.1 | NP_001073895.1 | 5 | 771 | ||
XM_005245603.3 | XP_005245660.1 | 6 | 934 | |||
XM_011510146.2 | XP_011508448.1 | 5 | 808 | |||
GenBank mRNA | BC115387.1 | - | 6 | 891 |