Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252288.3 | XP_005252345.1 | 13 | 1437 | ||
XM_011519150.2 | XP_011517452.1 | 13 | 1784 | |||
XM_011519151.2 | XP_011517453.1 | 13 | 1784 | |||
XM_017015243.1 | XP_016870732.1 | 13 | 1456 | |||
XM_017015245.1 | XP_016870734.1 | 13 | 1958 | |||
XM_017015246.1 | XP_016870735.1 | 13 | 1437 | |||
XM_017015249.1 | XP_016870738.1 | 12 | 1269 | |||
GenBank mRNA | AK225361.1 | - | 13 | 1373 | ||
EF611343.1 | - | 13 | 1388 |
Gene Symbol: CDC14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003595.1 | 1 | 1410 | |||
GenBank mRNA | BC028690.2 | - | 1 | 1398 | ||
BC068452.1 | - | 1 | 1409 |