Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005861.2 | NP_001005861.1 | 3 | 466 | ||
NM_002958.3 | NP_002949.2 | 3 | 466 | |||
GenBank mRNA | CN309980.1 | - | 3 | 310 | ||
S59184.1 | - | 3 | 499 | |||
X69970.1 | - | 3 | 466 | |||
X96588.1 | - | 4 | 371 |