Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024776.3 | NP_079052.2 | 5 | 2331 | ||
XM_005254670.2 | XP_005254727.1 | 8 | 2635 | |||
XM_005254671.2 | XP_005254728.1 | 7 | 2532 | |||
XM_005254673.2 | XP_005254730.1 | 5 | 2266 | |||
XM_005254674.2 | XP_005254731.1 | 6 | 2479 | |||
XM_005254675.4 | XP_005254732.1 | 2 | 1985 | |||
XM_011522033.2 | XP_011520335.1 | 9 | 2691 | |||
XM_011522034.2 | XP_011520336.1 | 10 | 2799 | |||
XM_011522035.2 | XP_011520337.1 | 12 | 2950 | |||
XM_011522036.2 | XP_011520338.1 | 11 | 2854 | |||
XM_011522037.2 | XP_011520339.1 | 11 | 2895 | |||
XM_011522038.2 | XP_011520340.1 | 10 | 2746 | |||
XM_011522039.2 | XP_011520341.1 | 11 | 2842 | |||
XM_011522040.1 | XP_011520342.1 | 9 | 2691 | |||
XM_011522041.1 | XP_011520343.1 | 9 | 2691 | |||
GenBank mRNA | AK091802.1 | - | 1 | 147 |