Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMTK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080434.1 | NP_001073903.1 | 12 | 3492 | ||
XM_011526411.2 | XP_011524713.1 | 12 | 3823 | |||
XM_011526412.2 | XP_011524714.1 | 12 | 3790 | |||
GenBank mRNA | AB067470.1 | - | 12 | 3465 | ||
BC036690.1 | - | 1 | 1088 |