Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 22 | 2143 | ||
XM_017020555.1 | XP_016876044.1 | 21 | 2151 | |||
XM_017020557.1 | XP_016876046.1 | 22 | 2234 | |||
GenBank mRNA | BC063885.1 | - | 22 | 2129 | 30383658 |