Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283.3 | NP_001274.1 | 2, 3 | 282 | ||
GenBank mRNA | AB015319.1 | - | 2, 3 | 199 | ||
AK312151.1 | - | 2, 3 | 248 | |||
BC003561.1 | - | 2, 3 | 201 | 3504050 | ||
BI597121.1 | - | 2, 3 | 287 | |||
BI598615.1 | - | 2, 3 | 287 | |||
BT006779.1 | - | 1, 2 | 172 |