Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSE1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256135.1 | NP_001243064.1 | 3 | 356 | ||
NM_001316.3 | NP_001307.2 | 3 | 356 | |||
XM_011528599.2 | XP_011526901.1 | 3 | 290 | |||
GenBank mRNA | AF053640.1 | - | 3 | 263 | ||
AF053641.1 | - | 3 | 290 | |||
AK297270.1 | - | 3 | 284 | |||
AK297314.1 | - | 2 | 189 | |||
AK307965.1 | - | 3 | 282 | |||
AK312306.1 | - | 3 | 287 | |||
BC108309.1 | - | 3 | 302 | 4540265 | ||
BC109313.2 | - | 3 | 269 | 40036129 | ||
BC109314.2 | - | 3 | 269 | 40036130 | ||
BG392946.1 | - | 3 | 301 | |||
DC402014.1 | - | 3 | 356 | |||
EF426455.1 | - | 2 | 167 | |||
U33286.1 | - | 3 | 290 |