Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005804.3 | NP_005795.2 | 3 | 433 | ||
NR_046366.2 | 3 | 433 | ||||
XM_006722606.3 | XP_006722669.1 | 3 | 373 | |||
XM_011527620.1 | XP_011525922.1 | 3 | 452 | |||
XM_011527621.2 | XP_011525923.1 | 2 | 319 | |||
GenBank mRNA | AB254849.1 | - | 2 | 315 | ||
AK293911.1 | - | 2 | 725 | |||
AK301847.1 | - | 4 | 516 | |||
BC001009.2 | - | 3 | 370 | 2820926 | ||
BC032128.2 | - | 3 | 379 | 5017293 | ||
DA432925.1 | - | 3 | 434 | |||
DB032777.1 | - | 4 | 526 | |||
U90426.1 | - | 3 | 410 |