Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005804.3 | NP_005795.2 | 6 | 780 | ||
NR_046366.2 | 6 | 780 | ||||
XM_006722606.3 | XP_006722669.1 | 7 | 905 | |||
XM_011527620.1 | XP_011525922.1 | 6 | 799 | |||
XM_011527621.2 | XP_011525923.1 | 6 | 724 | |||
GenBank mRNA | AB254849.1 | - | 5 | 662 | ||
AK301847.1 | - | 7 | 863 | |||
AL512698.1 | - | 1 | 644 | |||
BC001009.2 | - | 6 | 717 | 2820926 | ||
BC032128.2 | - | 6 | 726 | 5017293 | ||
U90426.1 | - | 6 | 757 |