Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX12P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002710.2 | 14 | 2421 | |||
NR_120453.1 | 13 | 1868 | ||||
GenBank mRNA | AL832768.1 | - | 14 | 2138 | ||
BC041851.1 | - | 16 | 2602 | |||
BU689660.1 | - | 1 | 351 |