Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321065.1 | NP_001307994.1 | 4 | 483 | ||
NM_001321066.1 | NP_001307995.1 | 5 | 545 | |||
NM_001645.4 | NP_001636.1 | 4 | 419 | |||
GenBank mRNA | AI310334.1 | - | 1 | 151 | ||
AI885116.1 | - | 1 | 141 | |||
AK225971.1 | - | 4 | 405 | |||
AK312036.1 | - | 4 | 300 | |||
BC009698.2 | - | 3 | 242 | 3889900 | ||
BC055093.1 | - | 5 | 471 | 6567705 | ||
BT007142.1 | - | 3 | 220 | |||
CA445083.1 | - | 1 | 166 | |||
CR456907.1 | - | 3 | 220 | |||
DA482306.1 | - | 5 | 545 | |||
M27359.1 | - | 3 | 178 | |||
X00570.1 | - | 3 | 276 |