Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000485.2 | NP_000476.1 | 1 | 53 | ||
NM_001030018.1 | NP_001025189.1 | 1 | 53 | |||
GenBank mRNA | BC106894.2 | - | 5 | 628 | ||
BC106895.2 | - | 5 | 631 | |||
BC107151.2 | - | 1 | 43 | 40032160 | ||
BM423481.1 | - | 77 | ||||
BM550173.1 | - | 1 | 76 | |||
CR749423.1 | - | 1 | 44 |