Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 5 | 666 | ||
NM_001318156.1 | NP_001305085.1 | 4 | 548 | |||
NM_001318157.1 | NP_001305086.1 | 5 | 984 | |||
NM_001318158.1 | NP_001305087.1 | 5 | 666 | |||
NR_134513.1 | 4 | 542 | ||||
NR_134514.1 | 4 | 542 | ||||
NR_134515.1 | 5 | 693 | ||||
XM_005251453.3 | XP_005251510.1 | 6 | 766 | |||
XM_006716765.3 | XP_006716828.1 | 4 | 427 | |||
XM_011517866.2 | XP_011516168.1 | 5 | 576 | |||
XM_011517867.2 | XP_011516169.1 | 3 | 206 | |||
XM_017014700.1 | XP_016870189.1 | 5 | 867 | |||
XM_017014701.1 | XP_016870190.1 | 4 | 537 | |||
XM_017014702.1 | XP_016870191.1 | 5 | 572 | |||
XM_017014703.1 | XP_016870192.1 | 4 | 325 | |||
XM_017014704.1 | XP_016870193.1 | 3 | 633 | |||
XM_017014705.1 | XP_016870194.1 | 3 | 303 | |||
XM_017014706.1 | XP_016870195.1 | 6 | 697 | |||
GenBank mRNA | AB006190.1 | - | 5 | 503 | ||
AK294028.1 | - | 5 | 477 | |||
AK297860.1 | - | 5 | 571 | |||
AK297885.1 | - | 5 | 800 | |||
AK300716.1 | - | 2 | 444 | |||
AK301949.1 | - | 4 | 505 | |||
AK310456.1 | - | 3 | 405 | |||
AK315879.1 | - | 2 | 435 | |||
BC062701.1 | - | 4 | 375 | 30324263 | ||
BC119672.1 | - | 4 | 374 | |||
BC119673.1 | - | 4 | 374 |