Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 8 | 1122 | ||
NM_001318157.1 | NP_001305086.1 | 7 | 1798 | |||
NM_001318158.1 | NP_001305087.1 | 7 | 1480 | |||
NR_134513.1 | 5 | 1237 | ||||
NR_134514.1 | 6 | 1356 | ||||
NR_134515.1 | 7 | 1507 | ||||
XM_005251453.3 | XP_005251510.1 | 9 | 1222 | |||
XM_006716765.3 | XP_006716828.1 | 7 | 883 | |||
XM_011517866.2 | XP_011516168.1 | 8 | 1032 | |||
XM_011517867.2 | XP_011516169.1 | 6 | 662 | |||
XM_017014700.1 | XP_016870189.1 | 8 | 1323 | |||
XM_017014701.1 | XP_016870190.1 | 7 | 993 | |||
XM_017014702.1 | XP_016870191.1 | 8 | 1028 | |||
XM_017014703.1 | XP_016870192.1 | 7 | 781 | |||
XM_017014704.1 | XP_016870193.1 | 6 | 1089 | |||
XM_017014705.1 | XP_016870194.1 | 6 | 759 | |||
XM_017014706.1 | XP_016870195.1 | 8 | 1511 | |||
GenBank mRNA | AB006190.1 | - | 8 | 959 | ||
BC119672.1 | - | 6 | 1188 | |||
BC119673.1 | - | 6 | 1188 | |||
DQ786216.1 | - | 3 | 334 |