Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001661.3 | NP_001652.2 | 2 | 437 | ||
XM_011524782.2 | XP_011523084.1 | 2 | 365 | |||
GenBank mRNA | AB060692.1 | - | 2 | 413 | ||
AK314951.1 | - | 2 | 429 | |||
BC000043.2 | - | 2 | 417 | 3509618 | ||
BI547763.1 | - | 2 | 468 | |||
L38490.1 | - | 2 | 412 |