Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181575.4 | NP_853553.1 | 10 | 1310 | ||
NR_126510.1 | 10 | 1310 | ||||
NR_126511.1 | 9 | 1506 | ||||
XM_005264392.3 | XP_005264449.1 | 9 | 1255 | |||
GenBank mRNA | AF100746.1 | - | 10 | 1083 | ||
AF100753.1 | - | 10 | 1110 | |||
AF100754.1 | - | 9 | 1308 | |||
AF165515.1 | - | 10 | 1090 | |||
AK023983.1 | - | 10 | 1088 | |||
AK055566.1 | - | 6 | 1942 | |||
AK293121.1 | - | 10 | 1129 | |||
AK304886.1 | - | 9 | 1252 | |||
AK308231.1 | - | 9 | 1309 | |||
AK310159.1 | - | 9 | 1308 | |||
BC001658.1 | - | 10 | 1173 | 3050204 | ||
BC033646.2 | - | 10 | 1125 | |||
CR749661.1 | - | 9 | 1181 |