Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCYRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001568.1 | 1 | 67 |
Gene Symbol: ARMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715573.2 | XP_006715636.1 | 2 | 201 | ||
XM_011536166.1 | XP_011534468.1 | 2 | 186 |
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG251503.1 | - | 1 | 492 |
Gene Symbol: COL19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535437.2 | XP_011533739.1 | 51 | 10573 | ||
XM_011535438.2 | XP_011533740.1 | 51 | 10301 | |||
XM_017010253.1 | XP_016865742.1 | 50 | 10531 | |||
XM_017010254.1 | XP_016865743.1 | 50 | 10519 | |||
XM_017010255.1 | XP_016865744.1 | 51 | 10512 | |||
XM_017010256.1 | XP_016865745.1 | 50 | 10264 | |||
XM_017010257.1 | XP_016865746.1 | 50 | 10447 | |||
XM_017010258.1 | XP_016865747.1 | 50 | 10180 |
Gene Symbol: LOC101930049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL700839.1 | - | 2 | 137 |
Gene Symbol: PCAT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024259.1 | 2 | 1467 | |||
GenBank mRNA | AK056805.1 | - | 2 | 1467 |
Gene Symbol: SNAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003082.3 | NP_003073.1 | 10 | 2153 | ||
GenBank mRNA | BC014984.1 | - | 10 | 2061 | 4872279 | |
BC019038.2 | - | 10 | 2058 | 4590014 |
Gene Symbol: ZFP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304358.1 | NP_001291287.1 | 3 | 2140 | ||
NM_174900.4 | NP_777560.2 | 4 | 2306 | |||
GenBank mRNA | AK056719.1 | - | 3 | 2143 | ||
AY429593.1 | - | 4 | 2309 | |||
BE301754.1 | - | 1 | 337 |
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647182.1 | - | 2 | 670 |
Gene Symbol: ZNF785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545753.2 | XP_011544055.1 | 4 | 2664 | ||
XM_017022967.1 | XP_016878456.1 | 4 | 2625 | |||
GenBank mRNA | AF086220.1 | - | 1 | 623 |