Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032515.4 | NP_115904.1 | 5 | 895 | ||
XM_011511696.2 | XP_011509998.1 | 5 | 699 | |||
XM_017004775.1 | XP_016860264.1 | 6 | 997 | |||
XM_017004776.1 | XP_016860265.1 | 6 | 923 | |||
GenBank mRNA | AF174487.1 | - | 5 | 744 | ||
BC006203.2 | - | 5 | 719 | |||
BT007272.1 | - | 4 | 603 | |||
BX460019.2 | - | 5 | 898 | |||
CN279175.1 | - | 2 | 188 |