Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322243.1 | NP_001309172.1 | 5 | 843 | ||
NM_001728.3 | NP_001719.2 | 6 | 1191 | |||
NM_198589.2 | NP_940991.1 | 5 | 843 | |||
NM_198590.2 | NP_940992.1 | 5 | 898 | |||
NM_198591.2 | NP_940993.1 | 4 | 741 | |||
XM_017027173.1 | XP_016882662.1 | 6 | 1080 | |||
GenBank mRNA | AB072923.1 | - | 5 | 676 | ||
AB085790.1 | - | 5 | 701 | |||
AB091680.1 | - | 5 | 671 | |||
AF548371.1 | - | 6 | 1070 | |||
AK095912.1 | - | 4 | 1259 | |||
AK296858.1 | - | 3 | 356 | |||
AK297880.1 | - | 6 | 1049 | |||
AK302231.1 | - | 5 | 711 | |||
AY358113.1 | - | 6 | 1062 | |||
BC009040.2 | - | 5 | 706 | 3867352 | ||
D45131.1 | - | 5 | 691 | |||
GU557064.1 | - | 5 | 865 | |||
GU557065.1 | - | 4 | 708 | |||
L10240.1 | - | 5 | 676 | |||
L20471.1 | - | 5 | 707 | |||
M87879.1 | - | 5 | 684 | |||
X64364.1 | - | 5 | 719 |