Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322243.1 | NP_001309172.1 | 5 | 651 | ||
NM_001728.3 | NP_001719.2 | 6 | 999 | |||
NM_198589.2 | NP_940991.1 | 5 | 651 | |||
NM_198590.2 | NP_940992.1 | 5 | 706 | |||
NM_198591.2 | NP_940993.1 | 4 | 549 | |||
XM_017027173.1 | XP_016882662.1 | 6 | 888 | |||
GenBank mRNA | AB072923.1 | - | 5 | 484 | ||
AB085790.1 | - | 5 | 509 | |||
AB091680.1 | - | 5 | 479 | |||
AF548371.1 | - | 6 | 878 | |||
AK095912.1 | - | 4 | 1067 | |||
AK297880.1 | - | 6 | 857 | |||
AK302231.1 | - | 5 | 519 | |||
AY358113.1 | - | 6 | 870 | |||
BC009040.2 | - | 5 | 514 | 3867352 | ||
BG480530.1 | - | 4 | 483 | |||
BI464857.1 | - | 5 | 575 | |||
D45131.1 | - | 5 | 499 | |||
GU557064.1 | - | 5 | 673 | |||
GU557065.1 | - | 4 | 516 | |||
L10240.1 | - | 5 | 484 | |||
L20471.1 | - | 5 | 515 | |||
M87879.1 | - | 5 | 492 | |||
X64364.1 | - | 5 | 527 |