Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000606.2 | NP_000597.2 | 4 | 515 | ||
GenBank mRNA | AW663384.1 | - | 3, 4 | 370 | ||
BC113624.1 | - | 4 | 490 | 8322676 | ||
BC113626.1 | - | 4 | 490 | 8322678 | ||
BX282392.1 | - | 3 | 253 | |||
M17263.1 | - | 4 | 466 | |||
M17999.1 | - | 4 | 501 | |||
X06465.1 | - | 4 | 501 |