Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014435.1 | NP_001014435.1 | 5 | 759 | ||
NM_005182.2 | NP_005173.1 | 5 | 607 | |||
XM_005256136.3 | XP_005256193.1 | 5 | 592 | |||
XM_011523311.1 | XP_011521613.1 | 5 | 601 | |||
XM_011523312.1 | XP_011521614.1 | 6 | 680 | |||
GenBank mRNA | AY075019.1 | - | 5 | 614 | ||
AY075020.1 | - | 5 | 761 | |||
BC033865.1 | - | 5 | 563 | 5185069 | ||
BI752037.1 | - | 5 | 741 |