Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFA2T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005187.5 | NP_005178.4 | 10 | 1737 | ||
NM_175931.2 | NP_787127.1 | 9 | 1403 | |||
XM_005256323.4 | XP_005256380.1 | 9 | 1715 | |||
XM_011523419.2 | XP_011521721.1 | 10 | 1839 | |||
GenBank mRNA | AB010419.1 | - | 10 | 1606 | ||
AB010420.1 | - | 9 | 1403 | |||
AK090985.1 | - | 1 | 415 | |||
AK314766.1 | - | 10 | 1592 | |||
BC047019.1 | - | 10 | 1587 | 5227349 | ||
BC062624.1 | - | 10 | 1607 | 5202447 |