Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001783.3 | NP_001774.1 | 3 | 607 | ||
NM_021601.3 | NP_067612.1 | 3 | 493 | |||
GenBank mRNA | AF283770.3 | - | 3 | 450 | ||
AK223371.1 | - | 3 | 605 | |||
AK315337.1 | - | 3 | 447 | |||
BC113731.1 | - | 3 | 442 | 8322783 | ||
BC113733.1 | - | 3 | 442 | 8322785 | ||
M74721.1 | - | 3 | 422 | |||
M80462.1 | - | 3 | 458 | |||
M86921.1 | - | 3 | 455 | |||
S46706.1 | - | 3 | 429 | |||
S75217.1 | - | 3 | 422 | |||
S79248.1 | - | 3 | 308 | |||
X83540.1 | - | 3 | 308 |