Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 6 | 680 | ||
AK223210.1 | - | 6 | 685 | |||
AK313583.1 | - | 6 | 677 | |||
AX775815.1 | - | 6 | 676 | |||
AX775817.1 | - | 6 | 673 | |||
BC002975.1 | - | 6 | 660 | 3542354 | ||
BC032651.1 | - | 6 | 643 | 5504755 | ||
M80461.1 | - | 6 | 623 | |||
M89957.1 | - | 6 | 701 | |||
S52229.1 | - | 6 | 607 | |||
S79249.1 | - | 5 | 295 | |||
X83539.1 | - | 5 | 295 |