Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004936.3 | NP_004927.2 | 1 | 451 | ||
NM_078487.2 | NP_511042.1 | 1 | 451 | |||
GenBank mRNA | AF004819.1 | - | 1 | 410 | ||
BC014469.1 | - | 1 | 419 | 4871014 | ||
BI560960.1 | - | 1 | 504 | |||
CR536529.1 | - | 1 | 91 | |||
JQ694046.1 | - | 1 | 239 | |||
JQ694047.1 | - | 1 | 239 | |||
JQ694048.1 | - | 1 | 239 | |||
JX391994.1 | - | 1 | 115 | |||
U17075.1 | - | 1 | 412 |