Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 4, 5 | 652 | ||
NM_001815.4 | NP_001806.2 | 5, 6 | 705 | |||
NR_102333.2 | 6, 7 | 1209 | ||||
GenBank mRNA | BC106728.1 | - | 5, 6 | 643 | 40015072 | |
D90277.1 | - | 5, 6 | 691 | |||
D90278.1 | - | 6, 7 | 1182 | |||
E03348.1 | - | 5, 6 | 691 | |||
E03349.1 | - | 7, 8 | 1064 | |||
L00692.1 | - | 5, 6 | 671 | |||
L00693.1 | - | 4, 5 | 618 |