Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 8 | 1234 | ||
XM_017020951.1 | XP_016876440.1 | 7 | 1280 | |||
GenBank mRNA | AK290101.1 | - | 8 | 1205 | ||
AK298198.1 | - | 8 | 1103 | |||
AK312282.1 | - | 8 | 1206 | |||
BC001190.1 | - | 8 | 1193 | 3354679 | ||
BC004914.2 | - | 8 | 1185 | 3537262 | ||
BC008323.1 | - | 8 | 1191 | 3358085 | ||
BC010002.2 | - | 8 | 1200 | 4135106 | ||
BC019259.1 | - | 8 | 1189 | 3503057 | ||
BC019281.1 | - | 8 | 1199 | 2823530 | ||
BM684665.1 | - | 1 | 238 | |||
CR542268.1 | - | 7 | 1128 | |||
EU794629.1 | - | 8 | 1189 | |||
FJ224337.1 | - | 7 | 1128 | |||
KU177977.1 | - | 7 | 1128 | |||
KU177978.1 | - | 6 | 1200 | |||
L47647.1 | - | 8 | 1171 | |||
M16364.1 | - | 7 | 1136 | |||
M16451.1 | - | 7 | 1151 |