Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080679.2 | NP_542410.2 | 56 | 4150 | ||
NM_080680.2 | NP_542411.2 | 59 | 4471 | |||
NM_080681.2 | NP_542412.2 | 57 | 4213 | |||
XM_011514299.2 | XP_011512601.1 | 54 | 3762 | |||
XM_011514300.2 | XP_011512602.1 | 53 | 3582 | |||
XM_011514302.2 | XP_011512604.1 | 52 | 3783 | |||
XM_017010250.1 | XP_016865739.1 | 60 | 4736 | |||
XM_017010251.1 | XP_016865740.1 | 52 | 3600 | |||
GenBank mRNA | J04974.1 | - | 32 | 2056 |