Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080679.2 | NP_542410.2 | 60 | 4538 | ||
NM_080680.2 | NP_542411.2 | 63 | 4859 | |||
NM_080681.2 | NP_542412.2 | 61 | 4601 | |||
XM_011514299.2 | XP_011512601.1 | 58 | 4150 | |||
XM_011514300.2 | XP_011512602.1 | 57 | 3970 | |||
XM_011514302.2 | XP_011512604.1 | 56 | 4171 | |||
XM_017010250.1 | XP_016865739.1 | 64 | 5124 | |||
XM_017010251.1 | XP_016865740.1 | 56 | 3988 | |||
GenBank mRNA | AK130938.1 | - | 6 | 422 | ||
J04974.1 | - | 36 | 2444 |