Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004074.2 | NP_004065.1 | 1 | 155 | ||
GenBank mRNA | AK311934.1 | - | 1 | 112 | ||
BC000501.1 | - | 148 | ||||
BC063025.1 | - | 1 | 155 | 6018884 | ||
BT009803.1 | - | 1 | 81 | |||
CR407687.1 | - | 1 | 81 |